WiFi Compact Microscopes, National Optical & Scientific Instruments
Supplier: National Optical
DCX5-205-LED
470337-676EA 592 USD
470337-676
WiFi Compact Microscopes, National Optical & Scientific Instruments
Microscopes
Ideal for hobbyists or for teaching microscopy to primary levels.
  • Easy grip design
  • Low-position focusing controls
  • Inward facing nosepiece
  • Cordless LED illumination
National is proud to introduce their own series of WiFi digital microscopes. The DCX5-205LED represents the latest in educational technology. With included Moticam X5, this WiFi digital microscope transmits live images to iOS or Android devices.
The DCX5-205LED features high-speed, full resolution imaging technology built into one of our most popular microscopes. Use it as a conventional microscope or share live images with your class using WiFi tablets or wireless laptops. High-speed, high-resolution to fit any budget.