Moticam X5 Plus, Digital WiFi Microscope Camera
Supplier: National Optical
D-MOTICAM X5
470337-612EA 422 USD
470337-612
Moticam X5 Plus, Digital WiFi Microscope Camera
Cameras
Turn any standard microscope into a digital microscope!
  • Generates own WiFi signal
  • Compatible with OS, IOS and Android devices
  • Industry standard C-mount design
  • Includes Motic Images software CD
Unlock the power of your microscope with the Moticam X5 Plus! Stream HD images to your WiFi enabled tablet, cell phone or computer. Connect up to five devices without the need for a router. Download the free MotiConnect app from the App store or the Google Play store.
Delivery information: Supplied with software, calibration slide, USB cable (1,8 m), and eyepiece adapters for binocular and monocular microscopes (30 and 30,5 mm diameter).