Biotechnology Modeling Kit
Supplier: 3-D Molecular Designs
CSBTK-01
470337-145CS 654 USD
470337-145
Biotechnology Modeling Kit
Educational Materials Biology Educational Materials AP Biology Learning Activities
Explore biotechnology topics
  • Interactive modeling
  • Covers PCR, restriction enzymes and Sanger sequencing
  • Reusable
Help your students achieve a better understanding of multiple biotechnology concepts including diagnostics, forensics and bacterial transformation with a single hands-on modeling kit!
Using the color-coded foam nucleotides in the Biotechnology Kit© and teacher-developed activities, students:
Model polymerase chain reaction and how flanking primers result in the exponential amplification of a short product.
Model the Sanger DNA sequencing method using chain-terminating fluorescent dideoxy nucleotide analogs.
Model DNA cloning using restriction endonucleases to cut DNA at specific sequences.
And more!