Chromosome Connections
Supplier: 3-D Molecular Designs
CSCHK-01
470337-144CS 474.55 USD
470337-144
Chromosome Connections
Educational Materials Biology Educational Materials AP Biology Learning Activities
Explore chromosome interactions and arrangements
  • Interactive modeling
  • Examine chromosomes at microscopic or molecular levels
  • Reusable
Help your students make the connection between chromosomes at the microscopic scale and chromosomes at the molecular scale! The Chromosome connections kit will enhance your classroom exploration of cell cycle, cell division and genetics.
Students can manipulate the colorful foam pieces in the kit to:
Model chromosome structure and anatomy.
Construct Punnett squares to connect inheritance of traits to chromosomes at the molecular level.
Compare and contrast mitosis and meiosis.
Model mechanisms that contribute to genetic variation.
Model chromosomal aberrations such as nondisjunctions, translocations, inversions, deletions and duplications.
And much more!