Flow of Genetic Information
Supplier: 3-D Molecular Designs
CSFGIK-01
470337-143CS 794.35 USD
470337-143
Flow of Genetic Information
Educational Materials Biology Educational Materials AP Biology Learning Activities
How does DNA function?
  • Interactive Modeling
  • Replication , Transcription And Translation Modeling
  • Reusable
Don't be surprised if one of your students blurts "Now I get it!" while using our Flow of Genetic Information Kit©. Manipulating the kit's color-coded foam nucleotides and placemats, students will model replication of the leading and lagging strands of DNA, model transcription as they copy one strand of DNA into mRNA using an RNA polymerase and model translation/protein synthesis as they decode the mRNA into protein on the ribosome placemat.