Ergotron K-Slot Lock Cable
Supplier: GLOBAL INDUSTRIAL COMPANY
WGB2308607
470336-502EA 118.75 USD
470336-502
Ergotron K-Slot Lock Cable
Security Locks
This K-slot (Kensington-style) keyed lock will secure a balance or other K-Slot equipped equipment.
  • K-Slot compatible
  • Keyed lock
  • Includes lock, cable, allen wrench, and 2 keys
  • 5-year warranty
Keep valuables safe with this K-Slot cable lock. Compatible with any piece of equipment which features a K-Slot for attachment (balances, computers, monitors, etc). Uses keys.