Reusable Silica Gel Desiccator Cartridge
Supplier: Bel-Art
F42049-0100
470336-462PK 100.35 USD
470336-462
Reusable Silica Gel Desiccator Cartridge
Desiccators Desiccants
Useful anywhere desiccation is needed, these 72 mm cartridges contain inert, non-corrosive, and non-toxic silica gel.
  • Clear polypropylene dish allows instant visual inspection of silica gel, which changes in color from blue to pale pink as it absorbs moisture
  • Highly porous mesh membrane covers the dish providing excellent moisture absorption
  • Desiccant can be easily regenerated by heating cartridges in a 50 °C (122 ºF) oven overnight or at 100 °C (212 °F) for 6 hours
  • For more rapid drying, remove the membrane lid, transfer the spent gel crystals into a glass or porcelain dish, and place in a 150 °C (300 °F) oven for one hour
  • Individually foil sealed
These cartridges also fit 'Space Saver' desiccators, in addition to other plastic desiccators of similar size.
Caution: May cause cancer and reproductive harm.