Desiccators, Vacuum, Clear Base, PP/PC
Supplier: United Scientific Supplies
55214 55215 55216 55217
470336-384EA 71.75 USD
470336-384 470336-386 470336-388 470336-390
Desiccators, Vacuum, Clear Base, PP/PC
Desiccators Desiccators, Plastic
These autoclavable desiccators are made of polypropylene (PP) and polycarbonate (PC).
  • Can hold a vacuum up to 740 mm Hg for 24 hours without any greasing
  • Autoclavable at 120 °C for 15 minutes
  • The transparent PC bottom provides increased viewing clarity
  • An internal groove on the flange holds the silicon rubber O-ring
  • Includes a perforated PP desiccator plate
  • The knurled knob on the top allows easy handling of the dome
  • The top dome, molded in rigid and transparent PC, gives a crystal-clear view of the sample
The PP stopcock has a self-lubricating PTFE plug. The three-way plug opens to create a vacuum, closes to hold a vacuum, and opens again to release a vacuum.