HOBO® Data Loggers Self-Describing 6' Pipe Temperature Sensor
Supplier: ONSET COMPUTER CORPORATION SE
SD-TEMP-P-06
470336-316EA 94.96 USD
470336-316
HOBO® Data Loggers Self-Describing 6' Pipe Temperature Sensor
Data Loggers
Temperature sensor with Self-Describing technology for quick, easy set up.
  • Available in 1.8 m (6')
  • Faster response time
  • Suitable for pipe mounting
Locking jack to ensure sensor is connected and stays connected.