Obliterase RNase and DNase Eliminator
Supplier: ALDON CORP SE
IS5603
470336-186EA 41.2 USD
470336-186
Obliterase RNase and DNase Eliminator
Cleaning Agents Decontaminants
Use Innovating Science’s Obliterase to eliminate RNase, DNase, and DNA contamination.
  • Use on glass, plastics, and stainless steel
  • Decontaminate beakers, flasks, pipettes, and tubes
  • Residue free
  • Phosphate free/biodegradable
Use full strength, do not dilute. Wet item thoroughly by spraying solution and ensure all surfaces are coated. Let item sit for a couple seconds. Let drain and rinse twice with DNase-free water. You can also soak the object in Obliterase, drain, and rinse twice with DNase-free water.