Electrolysis of Tin
Supplier: Ward's Science
IS7048
470336-162EA 24.8 USD
470336-162
Electrolysis of Tin
Educational Materials Chemistry Educational Materials Electrochemistry Learning Activities
Perform an electrolytic redox reaction to create tin crystals in a solution.
  • Understand and demonstrate electrolysis redox reactions
  • Aligned to NGSS practices
The electrolysis of a tin (II) chloride solution demonstrates the oxidation and reduction of Sn2+ ions as beautiful tin crystals grow and spread throughout the solution. There are enough materials to complete the demonstration five times.
Caution: WARNING: This product can expose you to chemicals including Phthalates, which is known to the State of California to cause reproductive harm.  For more information go to www.P65Warnings.ca.gov.