Digital Voltmeter, 0 - 20 V DC, Eisco Scientific
Supplier: Wards
PH1102A
470336-122EA 46.6 USD
470336-122
Digital Voltmeter, 0 - 20 V DC, Eisco Scientific
Electrical Meters Voltmeters
Light and portable digital voltmeter used to measure the potential difference between two points.
  • Light and portable with large LCD screen
  • 0 - 20 V DC, accuracy ±1 digit
  • Required 9 V battery (included)
Easy to read digital voltmeter is battery operated and can be used in the classroom, lab, or in the field. Designed to measure the potential difference between two points. Standard instrument for physics and electronic courses.