Ward's® Bowens Reaction Series Collection
Supplier: Avantor
470335-428KT 29.1 USD
470335-428
Ward's® Bowens Reaction Series Collection
Specimens Mineral Specimens Mineral Collections
Specimens illustrate important stages of mineral formation.
Samples included in the collection are representative of Bowen's Reaction Series, from both the continuous and discontinuous branches of minerals formed from the cooling of magma. This set is housed in a compartmented collection box. A chart detailing the reaction series and an identifrication list is included.
Contains 15 numbered samples, approximately 1×1" in size each.
Minerals: Albite, anorthite, augite, biotite, bytownite, enstatite, hornblende, labradorite, muscovite, oligoclase, olivine, orthoclase (2), phlogopite, quartz.