Sprint™ 8 Plus Clinical Centrifuge
Supplier: Ward's
Z207-A-CMB
470334-150EA 2960.5 USD
470334-150
Sprint™ 8 Plus Clinical Centrifuge
Centrifuges Benchtop Centrifuges
The Sprint™ 8 PLUS allows for the separation of serum or plasma in as little as 2 to 3 minutes.
  • Unique EZ-Scroll™ touch pad
  • Ideal for rapid production of serum and plasma
  • Program storage up to 99
  • Adjustable speed and timer
  • 8 Position fixed angle rotor rotor
  • Digital speed control in rpm or rcf with adjustable acceleration and deceleration rates (up to 6800 rpm/4445 ×g)
Additional features includes, A large back-lit, liquid crystal display, High capacity rotor, 8 tubes up to 15 ml (conical or round bottom), Timer up to 99 hr 99 mins, Storage of commonly used run-times, Control of speed in in both rpm and rcf, Adjustable acceleration and deceleration levels, Safety imbalance detection system, Momentary operation and Electrical lid lock.
Speed increment - 10 rpm or 10 rcf