BenchBlotter™ Fixed Speed Platform Rocker
Supplier: Ward's
BR1000
470334-082EA 545.4 USD
470334-082
BenchBlotter™ Fixed Speed Platform Rocker
Shakers and Mixers Rotating/Rocking Shakers
The BenchBlotter is an economical 'workhorse' rocker, designed for common molecular lab applications such as blotting, staining and destaining.
  • Optimized for blot washing and gel staining
  • Cold room and incubator safe
  • Dual platform accessory doubles work surface
  • Gentle mixing, 12 rpm ±8° tilt
There are no complicated controls, no settings or adjustments to be made, just press the power switch and start rocking.