Room Divider, 6' Height, 7 Sections, PET, National Public Seating
Supplier: National Public Seating
RDB6-7PT02 RDB6-7PT04 RDB6-7PT10
470332-900EA 2717.15 USD
470332-900 470332-908 470332-916
Room Divider, 6' Height, 7 Sections, PET, National Public Seating
Prefabricated Rooms
Divide your multipurpose room with portable Robo™ dividers made with 100% recycled materials.
  • Constructed from 100% recycled environmentally friendly PET materials
  • Panels are sound dampening with a noise reduction coefficient of 0.84
  • Flame retardant: B1 grade
  • Panels are tackable
  • Self-leveling casters
  • Full-length hinges
  • Heavyweight- non tipping
Divide your multipurpose room, medical center or house of worship with the ultimate partition: the durable, flexible and portable Robo™ dividers. Room to room transportation is made simple with full length handles and spring enhanced casters that glide right over thresholds and uneven flooring. They feature Tech-Lock brakes at both ends for a secure stop. Our portable dividers are made with steel, aluminum-framed panels and can be ganged together with magnet connectors built into the divider ends. Dividers are available in PET and clear acrylic.
They stand 6' tall and come in four lengths. Go green with the environmentally-friendly PET divider, made with 100% recycled materials. The PET panels dampen sound, so outside distractions won't stand a chance. Or go with our clear acrylic Robo™ dividers to see and hear it all while maintaining a distance. The acrylic panels are crafted with lightweight, non-yellowing OPTIX® acrylic, a highly durable material that resists intense force or shock. Robo™ - Robust. Easy. Portable.
Certifications: ANSI-BIFMA Certified and MAS Certified Green.
Ordering information: Two-week Quickship. Ten-year warranty. Clear acrylic dividers are available separately.
Delivery information: Shipped fully assembled.