Updated Wimshurst Machine, United Scientific
Supplier: United Scientific Supplies
WIMS02
470332-762EA 230.3 USD
470332-762
Updated Wimshurst Machine, United Scientific
Educational Materials Physics Educational Materials Electricity/Magnetism Learning Activities
A superior quality Wimshurst machine for demonstrating high voltage electric charges.
  • Demonstrate the build up of charge
  • Vividly demonstrate the strength of a stored charge and the distance for to jump to ground
This unit features acrylic rotating disks, good quality foil strips and brushes, insulated cylindrical capacitors, and a durable plastic platform. Guaranteed to produce large sparks for easy classroom demonstration.