United Scientific Supplies Well Plate, 12-Well, Sterile
Supplier: United Scientific Supplies
UNWPLATE-12-PK/10
470332-698PK 20.85 USD
470332-698
United Scientific Supplies Well Plate, 12-Well, Sterile
Microplates Deep Well Microplates
Clear plastic plate features deep uniform wells and include lids.
  • Sterile and individually wrapped
  • Each well has an alpha-numeric marking
The 12-well plate measures 5" long×3-¹/₄" wide×⁷/₈" high. Flat-bottomed wells are ³/₄" deep and approximately 25 mm in Ø.
Packaging: Sold in packs of 10.