United Scientific Supplies Dropper Bottle, Unassembled, LDPE, 15 ml
Supplier: United Scientific Supplies
PDB015-U-PK/24
470332-692PK 9.6 USD
470332-692
United Scientific Supplies Dropper Bottle, Unassembled, LDPE, 15 ml
Bottles Dropping and Dispensing Bottles
This dropper bottle is made of translucent and low density polyethylene.
  • Bottle can be used with or without dropper
  • Comes in a pack of 24
For use when dispensing small drops of a solution. The drop sizes are uniform for consistent dispensing of liquids.