Safety Goggles, Clear, United Scientific
Supplier: United Scientific Supplies
UNGOGGLE-P
470332-656EA 2.3 USD
470332-656
Safety Goggles, Clear, United Scientific
Goggles
Goggles feature vents to prevent fogging and an adjustable band for a comfortable fit.
  • Comfortable head band
  • PVC lenses
Flexible safety goggles are manufactured from PVC and are designed to protect against splashes and impact. Four vents to prevent fogging and an adjustable band for a comfortable fit.
Certifications: Safety goggles are ANSI Z87.1-2020 certified.