Drop-Boy Dropper Bottle
Supplier: Dynalon
805224-0050
470332-512PK 66.15 USD
470332-512
Drop-Boy Dropper Bottle
Bottles Dropping and Dispensing Bottles
Precise drop dosing.
  • Bottle should have dropper tip cut off before first use
  • Reclosable using screw-on cap
Drop-Boy is a HDPE dropping bottle with a LDPE drop dispenser and separate polypropylene screw cap.