Forensic Investigation: Stages of Decomposition in Fetal Pigs
Supplier: Cyrgus
PK-69FI
470332-248KT 48.7 USD
470332-248
Forensic Investigation: Stages of Decomposition in Fetal Pigs
Specimens Preserved Specimens Preserved Animal Materials
In this activity, students will observe and study the process of decomposition using a fresh fetal pig.
  • Includes fresh pig and materials
  • Guide to the five stages of decomposition
  • Complete study of decomposition in under 4 weeks
Although admittedly not for the faint of heart, this activity brings the world of forensic science to life in an unfgorgettable way. Experience the process of decomposition first-hand with a real decomposing fetal pig. Requires an undisturbed outdoor area to lay out the fresh, fetal pig.
Number of setups: 1