Kartell® Pump Dispensing Bottle, Dynalon
Supplier: Dynalon
205135
470331-652CS 52.85 USD
470331-652
Kartell® Pump Dispensing Bottle, Dynalon
Bottles Dropping and Dispensing Bottles
New improved pump dispenser.
  • FDA compliant material
  • Pump dispenser suitable for liquids and creams
Supplied with polypropylene closure, this 500 ml pump dispensing bottle is highly suitable for dispensing liquids and creams. Wide surface area is easily labeled with contents. Each pump dispenses approximately 2.5 ml (0.0845 oz.), and a ¹/₄ turn locks the pump.
Suitable for use with food grade products.