VWR® Advanced Binocular Microscope
Supplier: VWR International
470331-644
470331-644EA 1248.4 USD
470331-644
VWR® Advanced Binocular Microscope
Microscopes
Offers the versatility and precision necessary for research and clinical applications.
  • Siedentopf binocular head
  • Parfocal and parcentered optics
  • Coaxial coarse and fine adjustment controls
  • Three-year manufacturer's warranty
Multiple features make this a practical choice for classroom and clinical environments. The combination of Cool-white LED lighting and glass-coated optics provide crisp and clear images allowing students to view more details and minute structures.
Delivery information: Microscope includes a manual and a dust cover.