VWR® Standard / VWR® Advanced Standard and Advanced Compound Microscopes
Supplier: VWR International
470331-632 470331-634 470331-636
470331-632EA 510.06 USD
470331-632 470331-634 470331-636
VWR® Standard / VWR® Advanced Standard and Advanced Compound Microscopes
Microscopes
Quality microscopes at an affordable price.
  • Parfocal and parcentered optics
  • Separate coarse and fine adjustment controls
  • Three-year manufacturer's warranty
These durable microscopes feature all-metal bodies and all-glass-coated optics. They provide crisp, clear images allowing students to view more details and minute structures.
Delivery information: All microscopes include a manual and a dust cover.