Edvotek Diagnosing Huntington's Using PCR
Supplier: Edvotek
1125
470331-034EA 231.3 USD
470331-034
Edvotek Diagnosing Huntington's Using PCR
Educational Materials Biology Educational Materials PCR Learning Activities
Detect genetic markers for a disease.
  • Perform standard DNA Fingerprinting techniques with real-world context
  • Combine mendelian genetics with medical diagnosis
In this experiment, students will conduct a DNA fingerprinting exercise on simulated patient samples to determine if family members are heterozygous or homozygous for Huntington's Disease. Students will then analyze the amplified DNA segments by agarose gel electrophoresis.
Additional items needed but not included: Thermal cycler, horizontal gel electrophoresis equipment, microcentrifuge, Light box (UV or White-light), micropipettes and tips, microwave or hot plate, 250 ml flasks or beakers, hot gloves, and disposable lab gloves.
Ordering information: Kit will ship immediately upon ordering. Please call to specify a future shipping date.
Caution: Some components require freezer storage.