Edvotek Cell Types in the Brain
Supplier: Edvotek
1110
470331-030EA 131.3 USD
470331-030
Edvotek Cell Types in the Brain
Educational Materials Biology Educational Materials Genetics Learning Activities
Examine the differences between cell types in the brain based on their profiles of proteins.
  • Includes lesson, study questions and answers
  • Perform SDS-PAGE electrophoresis of proteins in a real-world context
  • Precast polyacrylamide gels not included but available separately
The brain is an incredibly complex organ and is responsible for regulating almost everything within our body. It allows us to form complex thoughts, read, write, move, breathe, play sports, and listen to music. It does this through a network of cells working together to function.
Additional items needed but not included: 12% denaturing polyacrylamide gels (3), vertical gel electrophoresis equipment, shaker platform (optional), micropipettes with tips, microtest tubes, beakers, transfer pipets, graduated cylinders, plastic wrap, scissors, trays, forceps, several packs of paper towels, latex or vinyl lab gloves, safety goggles, methanol, and distilled water.
Ordering information: Kit will ship immediately upon ordering. Please call to specify a future shipping date.
Caution: Some components require freezer storage.