Robolink CoDrone Mini
Supplier: Klinger
RL-CDM-100
470330-994EA 170.85 USD
470330-994
Robolink CoDrone Mini
Educational Materials Physics Educational Materials Applied Physics Learning Activities
CoDrone Mini fits in the palm of your hand.
  • Perfect for ages 8+
  • Code in Blockly or Python
  • Compatible with Windows, macOS, and ChromeOS
  • 60 min battery charge
This is the CoDrone Mini. With its smaller propellers, the ability to flip, a rounded frame, and a lighter form factor, the CoDrone mini is a great starter drone and is perfect for younger audiences who are interested in getting into programming with drones.
We cover the basics starting from learning about loops and conditionals then going into much more advanced topics like reading sensor data and using it to graph data in Python. Students get tutorials, teachers get lesson plans. Learning what functions and variables are is much more exciting when it makes your drone draw a star in the air or do a flip. Do a bunch of activities that keep moving while you learn.