DeskBrite™ 300 Magnifying Desk Lamp
Supplier: CARSON OPTICAL
LM-30
470330-750EA 46.8 USD
470330-750
DeskBrite™ 300 Magnifying Desk Lamp
Magnifiers
Versatile hands-free magnifier for crafts, hobby, and inspection.
  • Flexible goose neck
  • 2×/5× magnification
  • 4.3" lens
  • Four COB LEDs
The LM-30 DeskBrite™ 300 is a 2× Aspheric COB LED magnifier desk lamp with a 5× power spot lens. This desk lamp features four super-bright COB LED lights with a diffusive cover. Light diffusers work to eliminate LED hot spots, blending and evenly distributing the light.
COB LEDs are brighter than traditional LEDs, provide a greater uniformity of lights, and consume less power. The large 4.5" crystal-clear aspheric, acrylic lens provides a large viewing area for any hobby. Aspheric lens magnifiers allow for increased magnification and image clarity while greatly reducing edge to edge distortion.
The DeskBrite™ 300 LED Magnifier Desk Lamp uses USB-C to US wall adapter with 6-foot cord, or it can be powered by using 3 AA batteries (not included).