Spigot, LDPE
Supplier: Dynalon
612935
470330-666CS 194.4 USD
470330-666
Spigot, LDPE
Stopcocks
Standard drum spigot.
Plastic spigots are molded of low-density polyethylene of white with 3/4" IPS thread. Plastic spigots have no gaskets or inside threads to leak. Will fit standard threaded drums.