Goggle Sanitizing Cabinet
Supplier: Wards
GGSN10
470017-114EA 845.85 USD
470017-114 470330-526
Goggle Sanitizing Cabinet
Sterilizers UV Sterilisers
This cabinet quickly and safely sanitizes goggles for the entire classroom.
  • Capacity of 36 goggles
  • UV-lamp timer
  • Welded seams
  • Tight-fitting piano hinges
  • Double-latching doors
  • Replacement bulb is a Philips T8 18" 25w TUV germicidal fluorescent
  • Cord is 8 feet long
Provide your classroom with sanitized goggles in just minutes. With a capacity of 36 goggles, this sturdy, heavy-gauge steel goggle sterilizer features a UV lamp timer to ensure proper UV exposure and germ eradication. The unit will not operate unless cabinet doors are closed, eliminating the worry of UV exposure. In addition, the unit offers safety features such as welded seams and tight-fitting piano hinges to contain the UV light.
The indicator light tells users when the sterilization process is occurring, and double-latching doors ensure safety.
Certifications: CSA approved