Advantage Economy Indirect Vent Goggles
Supplier: DR INSTRUMENTS INC. SE
DR81210E
470330-406EA 12.25 USD
470330-406
Advantage Economy Indirect Vent Goggles
Goggles
Indirect vent and fog-free.
  • Lightweight
  • Green tinted
  • Adjustable headband
Sellstrom advantage indirect vent economy protective goggles are lightweight, green tinted PVC. The soft body gently conforms to the face, providing a secure and comfortable fit. UV absorbing polycarbonate lens provide ultimate protection. Adjustable 18" green elastic headband secures the goggles comfortably to the face.
Certifications: Meets ANSI Z87.1-2003 standards. CUL certified to meet CAN/CSA Z94.3 standards.