MyLab Simulation of COVID-19 Antibody Test
Supplier: Edvotek
1219
470329-946EA 71.7 USD
470329-946
MyLab Simulation of COVID-19 Antibody Test
Educational Classroom Kits and Activities
Teach students about COVID-19 diagnostic testing at home.
  • Single student kit ideal for remote learning
  • Understand current COVID-19 testing
  • No human products or viral particles are used
  • Includes study questions and answers
With this simulated ELISA kit, your students will be able to perform a simulated COVID-19 testing procedure right from their home. Students will learn about antibody-based detection techniques, ELISA and current techniques for COVID-19 testing.
User to provide: Distilled or deionized water, small cup, permanent marker, timer or clock, gloves and goggles.
Packaging: Store at room temperature.
Caution: Choking Hazard. Not appropriate for children under five years old. Kit does not contain SARS-CoV-2 virus, nor can it identify viral infection in patient samples.