Basic Brain Model Bundle
Supplier: Avantor
470329-750EA 497.79 USD
470329-750
Basic Brain Model Bundle
Models Human Biology Models
Bundle and save.
  • 3B Smart Anatomy included
  • Two models for basic brain anatomy
These two models are often purchased together. The basic two part brain model is ideal for learning general brain structure. Life sized, and divided on the medial plane, this brain model is also labeled on the model. This makes it easier to use for testing and review with structures identified by number. Like most brain models though, it does not show the ventricles. The ventricle model illustrates these structures that are missing from a general brain anatomy lesson. Ventricles from both sides are shown, and presented on a base. Combined, these two models nicely round out an introductory brain anatomy lesson.