Ward's® Simulating Urinalysis Kit
Supplier: Avantor
470190-606EA 112.99 USD
470190-606 470190-612 470329-604
Ward's® Simulating Urinalysis Kit
Educational Classroom Kits and Activities
What can we learn from our urine?
  • Kit Employs NGSS Life Science DCI LS1
  • Pre- and Post-Laboratory Assessments Included
  • Kit Aligned with National Standards
  • Assessments and Vocabulary Words Target Common Core Standards
  • Estimated Lab Time: 30-45 Minutes
In this lab, students test Ward's safe and nontoxic simulated urine samples for pH, sugar, and protein and observe the samples through a microscope for further analysis. The students are provided with four different case studies and simulated urine for each patient. They perform a urinalysis on each sample. Students learn the basis of urinalysis and how it is used to diagnose medical problems while they employ the basic Structures and Properties (LS1) Framework Principles.
This lab activity takes approximately 30 to 45 minutes to complete. It is aligned with all published National Standards. Pre- and post-laboratory assessments and vocabulary words target specific Science and Engineering Practices and Common Core standards. The teacher guide contains specific Did You Know concepts that link the real world with the scientific standards explored. This kit includes a student copymaster and material for 10 setups.
Certifications: Kit Employs NGSS Life Science DCI LS1.
Ordering information: This kit includes simulated urine samples, pH strips, Benedict’s solution, biuret solution, graduated plastic pipets, medicine cups, a package of microscope slides, a package of coverslips, and a urine specimen microscope slide. Test tubes, test tube holders, 400 mL beakers, a hot plate, a compound microscope, heat protective gloves, and water are needed but not included.
Delivery information: Due to the nature of this item, it cannot be shipped by air under any circumstances. Please select ground shipping at time of checkout.