Disposable 3-Ply Facemasks
Supplier: Wards
BI0368PK50
470329-602EA 45.65 USD
470329-602
Disposable 3-Ply Facemasks
Masks Hygiene Masks
Non-surgical facemasks.
  • Breathable
  • Stretchy, elastic ear loops
  • Excellent for general-purpose, everyday use
Disposable, non-surgical 3-ply face masks feature a melt-blown polypropylene filtration layer. Masks include three, non-woven layers for optimal comfort and a filter for added protection. It is recommended to replace masks every 3 to 5 hours. Always wear with the blue-side facing out and the nose strip at the top.
Individual masks measure 3.6×6.75".
Packaging: Masks are packaged in a box of 50.