Ward's® Biochemistry Of Life's Macromolecules Lab Activity
Supplier: Avantor
470154-042KT 119.29 USD
470154-042 470008-048 470329-248
Ward's® Biochemistry Of Life's Macromolecules Lab Activity
Educational Classroom Kits and Activities
Students research a variety of macromolecules.
  • Activity: Researching Macromolecules
  • Students Determine Properties of Proteins, Lipids, and More
  • Estimated Lab Time: 40 Minutes
  • Material for 10 Setups
  • Aligned to NGSS standards (HS: PS3, LS1)
Using indicator solutions, students record and observe the reactions of proteins, carbohydrates, lipids, and vitamin C to determine their properties.
This kit includes instructions and material for 10 setups.
Accessories information: A refill kit is available and contains only consumable component items. It does not contain additional student or teacher guides.
Ordering information: This kit includes ascorbic acid powder, iodine solution, vegetable oil, biuret solution, copper sulfate, sudan III, starch solution, microfuge trays, microfuge tubes, and pipets. DI water is needed but not supplied.
Delivery information: Due to the nature of this item, it cannot be shipped by air under any circumstances. Please select ground shipping at time of checkout.