3B Scientific® Advanced Wearable Auscultation and Standardized Patient Lab
Supplier: American 3B Scientific
8000998
470329-220KT 20500.09 USD
470329-220
3B Scientific® Advanced Wearable Auscultation and Standardized Patient Lab
Models Human Biology Models
Perfect for your classroom or clinical simulation center.
  • Models, charts and simulation
  • Includes 3B Smart Anatomy
Auscultation is an essential clinical skill needed to assess and monitor patients’ conditions. With the Advanced Wearable Auscultation Lab, teaching and learning this skill becomes even more versatile, economic and impactful.
Patient care simulation has long been required to sacrifice realism for accuracy, or vice versa. Using the Bionic Hybrid Simulator in the Advanced lab kit we have broken barriers to achieve both accurate and realistic simulation. Test students’ skills without giving away signs and symptoms. Witness their bedside interactions with a real Standardized Patient and watch their level of engagement grow as they build confidence in their clinical skills.