Omnimed® Mounting Magnets
Supplier: OMNIMED INCORPORATED SE
305202
470329-024PK 47.49 USD
470329-024
Omnimed® Mounting Magnets
Magnets
No more drilling!
  • 1.25" diameter magnets
  • Fits items with a mounting depth of up to 0.10"
Omnimed® mounting magnets are perfect for attaching glove boxes, wall pockets, or any item with standard screw mounting holes to a metal surface. Each magnet is rated to 16 lbs. of force to ensure a secure hold. Each magnet contains a threaded hole and a truss head screw. Simply put the screw through the mounting hole and tighten securely.
Packaging: Pack of 4.