Omnimed® Junior Heavyweight Infusion Stand
Supplier: OMNIMED INCORPORATED SE
741301
470328-986EA 362.99 USD
470328-986
Omnimed® Junior Heavyweight Infusion Stand
Support Stands
A heavyweight, four-hook IV pole.
Omnimed® Junior Heavyweight IV infusion stands have a stainless steel pole that has four rams-hooks capable of holding up to four 3000 CC containers. The stand manually rises to 7' and has an air glide system which controls the descent of the pole. The five-leg black powder-coated aluminum base has a 22.5" diameter and dual wheel casters for smooth and stable movement.