Stainless Steel Micro Lab Scoop with Spatula, Eisco
Supplier: Wards
CH0635D
470328-216EA 4.95 USD
470328-216
Stainless Steel Micro Lab Scoop with Spatula, Eisco
Spatulas
Features one scoop end and one flat spatula end. Designed for use with small samples. Perfect for a variety of laboratory uses.
  • Made of high quality polished stainless steel
  • Autoclavable, reusable
  • Feature one scoop end and one spatula end
  • Designed for use with small samples
Stainless steel lab scoop with spoon on one end and flat edge with on the other end. Total legnth is 5.9", Spatula measures 0.2" wide, scoop measures 0.2" wide. These scoops are great for adding small amounts of ingredients and mixing.