Polypropylene Lab Scoops with Spatula, Eisco
Supplier: Wards
CH0631APK10
470328-200EA 13.8 USD
470328-200
Polypropylene Lab Scoops with Spatula, Eisco
Spatulas
Features one scoop end and one spatula end with knife edge. Perfect for a variety of laboratory uses.
  • Made of high quality, chemical and heat resistant polypropylene
  • Autoclavable, reusable
  • Feature one scoop end and one spatula end with knife edge
Blue polypropylene lab scoop with a spoon on one end and flat edge with on the other end. Made of polypropylene which is autoclavable and excellent replacement of metallic scoops. Total Length 150 mm (5.90"), Oval scoop measures 19 mm (0.75") Wide and 26 mm (1.02") long, Flat side measures 20 mm (0.79") wide by 26 mm (1.02") long. Handle is 16 mm (0.63") wide, 3 mm (0.12") thick. These scoops are great for adding small amounts of ingredients and mixing. Pack of 10.