Innovating Science® Engineering a Drug Delivery System
Supplier: Ward's Science
IS3403
470328-172KT 71.5 USD
470328-172
Innovating Science® Engineering a Drug Delivery System
Educational Materials Biology Educational Materials Human Biology Learning Activities
Learn about drug delivery systems, which can control the transport and release of medicine in the body.
  • Students will gain an understanding of drug delivery and its importance by testing simulated delivery systems in multiple environments that mimic different parts of the body.
  • Aligned to NGSS Practices
  • Performance expectations: MS-ETS1-1, MS-ETS1-2, MS-LS1-3, HS-ETS1-2
  • Disciplinary core ideas: EST1.A, ETS1.B, LS1.A, ETS1.C
  • Cross cutting concepts include influence of science enginerring and technology on society and the natural world, systems and system models, science is a human endeavor
  • Engineering practices include asking questions and defining problems, engaging in argument from evidence, constructing explanations and designing solutions
Biomedical engineers design devices, equipment, and processes to improve medical treatment methods and the quality of human health and life. They apply engineering principles to biology and medicine to create products such as prosthetic limbs and artificial organs, among others. One major focus of biomedical engineers is the design of drug delivery systems, which can control the transport and release of medicine in the body to more effectively treat diseases and illnesses. In this experiment, students will gain an understanding of drug delivery and its importance by testing simulated delivery systems in multiple environments that mimic different parts of the body. The delivery systems will be analyzed and compared, allowing students to engineer an optimal solution and learn what properties must be considered when designing new drug delivery systems. This lab has enough materials for 15 groups. Teacher's manual and student guide included.
Delivery information: Kit includes instructional manual and SDS booklet.
Caution: WARNING: This product can expose you to chemicals including strong inorganic acid mists containing sulfuric acid, which are known to the state of California to cause cancer. For more information go to www.P65Warnings.ca.gov