Accucold Pharma-Vac Performance Series Refrigerators
Supplier: FELIX STORCH, INC./SUMMIT
ARG3PV
470327-958EA 3009 USD
470327-958 470327-960 470327-962 470327-964 470327-966 470327-968 470327-970 470327-972 470327-974 470327-976 470327-978 470327-980 76575-782 76575-784 76636-534 76636-542 76636-536 76636-518 76636-512 76636-538 76636-540 76636-532 76636-514 76636-516 76636-508 76636-510
Accucold Pharma-Vac Performance Series Refrigerators
Refrigerators
Designed and purpose-built for pharmacy, medication, and vaccine applications to support meeting CDC/VFC guidelines.
  • Solid and glass door refrigerators from 1 to 15 cubic feet
  • Built-in readout for current and high/low temperature
  • Audible and visual high/low temperature alarms
  • Optimized efficient performance for stable temperature
  • Eco-friendly natural refrigerants for a greener footprint
These Pharm-Vac Performance refrigerators are supplied as standard with an adjustable control range from 2 to 8 °C. The microprocessor temperature controller is externally located to minimize door openings, helping to protect stored contents from ambient temperature. The buffered temperature probe is encased in a glycol-filled bottle to better simulate temperature of the stored product, with an additional sensor that reads air temperature to ensure superior temperature control. The current and min./max. temperature display is viewable in Celsius or Fahrenheit to the nearest tenth of a degree. The audible and visual temperature alarm sounds if the unit's interior goes out of range. Additional alarms include a power failure alert, door open alarm, and sensor failure alarm. This unit is equipped with remote alarm contacts in the rear and a ½" probe port on the right hand side to accept additional monitoring devices.
Inside, they utilizes adjustable cycle defrost operation to avoid temperature spikes. Optimized forced air cooling with two internal fans ensures excellent temperature stability and uniformity with rapid recovery after door openings. The adjustable plastic-coated wire shelves can be positioned at ½" increments to accommodate virtually any sized item. We include seven shelves, more than competitor products providing to maximize interior shelving storage area. This unit includes LED internal lighting with an on/off rocker switch. The interior is constructed from white powder coated metal to ensure added durability and better temperature retention.
They come in a white cabinet finish with a blue accented control panel and easy-grip handle. The user-reversible double pane tempered glass door provides a full view of the interior. These models include a factory-installed lock and ships with two keys for staff use. Powerful magnetic gaskets ensure a positive seal.
Designed with hydrocarbon refrigerant, the Pharm-Fac Performance Refrigerators provide an energy efficient solution to vaccine storage. These units are Pyxis, Omnicell, and AcuDose RX compatible.
Delivery information: They are delivered with 100% guarantee, 2 year-parts and labor and 5 year compressor warranty.