Rare Earth Magnet
Supplier: PICOSOLUTIONS GROUP INC SE
PS-2016
470126-945EA 5.15 USD
470126-945 470326-954
Rare Earth Magnet
Magnets
Compatible with Variety of Windmill Kits
  • Strength 0.395 Tesla/3,950 Gauss
  • Size 1.875×0.875×0.375" thick
  • Pole marked with adhesive sticker
This ceramic, grade 5 magnet is ideal for use in constructing the alternator used in a variety of wind turbine kits. Use caution: ceramic magnets are magnetically strong, but physically fragile and can chip or crack easily.
Caution: WARNING! These magnets are strong and can pinch your fingers if not handled carefully. Persons wearing pacemakers should not handle strong magnets.