LaMotte® TesTab® Phosphate Test Kit
Supplier: LA MOTTE CHEMICAL PROD CO SE
5892
470326-584EA 39.1 USD
470326-584
LaMotte® TesTab® Phosphate Test Kit
Environmental Test Kits
Refill for The Comprehensive Water Monitoring Kit 470004-240 and LaMotte® TesTab® Water Investigation Kit 470007-270
  • 100 Tests
  • Range 0, 1, 2, 4 ppm
Each kit contains easy-to-read diagrammed instructions, a color chart, a test factor flash card and all the necessary apparatus and non-hazardous TesTabs® to perform 100 tests