Ward’s Pure Preserved Individual Dissections
Supplier: Avantor
470326-264EA 29.99 USD
470326-264 470326-266 470326-268 470326-270 470326-272 470326-274 470326-276 470326-278 470326-280 470326-282 470326-284 470326-286 470326-288 470326-290 470326-292 470326-294 470326-296 470326-298 470329-250 470330-112 470330-250 470330-252 470330-254 470330-256 470330-258 470330-260 470330-262 470330-264 470330-266 470330-268 470330-270 470330-272 470330-274 470330-276 470330-278 470330-280 470330-282 470330-284 470330-286 470330-288 470330-290 470330-292 470330-294 470330-296 470330-230 470330-232 470330-234 470330-236 470330-238 470330-240 470330-242 470330-244 470330-246 470330-248
Ward’s Pure Preserved Individual Dissections
Specimens Preserved Specimens Preserved Animal Materials
Everything you need for individual student dissections, whether at home or school.
  • Select individual specimens
  • Individual dissection accessory bundles includes everything needed to perform multiple dissections (Not available in Canada)
  • For specimens up to 9", select the small accessory bundle; for specimens above 9", select the medium accessory bundle
  • Digital dissection guides available for free with purchase
These dissection materials are perfect for remote learning or home schooling situations. Select desired specimens, and pair with the individual dissection accessory bundle to receive everything needed to perform multiple dissections. Specimens are Pure preserved specimens (unless otherwise indicated), which replaces the unpleasant formalin experience with a low odor solution that provides a uniquely safe, comfortable and high quality dissection experience.
Delivery information: The individual dissection accessory bundle includes 1 pair of goggles, 5 disposable aprons, 5 pairs of large disposable gloves, a Styrofoam tray, and a basic dissection instrument kit containing a scalpel, forceps, scissors, dropper, ruler and T-pins in a plastic pouch.
Small dissection bundle includes a 8×10" Styrofoam tray for smaller dissection specimens. Medium dissection bundle includes 12×16" for medium to large dissection specimens.