Health Smart® Basal Digital Thermometer
Supplier: HEALTH SMART SE
15-639-000
470326-222EA 16.65 USD
470326-222
Health Smart® Basal Digital Thermometer
Thermometers Digital Thermometers
Superior accuracy.
  • Includes case and tracking charts
  • Single button activation
Teach you students about fertility cycles. A Basal thermometer is designed to display readings to a 100th of a degree. This allows for a very precise measurement of body temperature. When used correctly it can be an indication of ovulation dates.