Bottles with Dropper, Boston Round, Amber, United Scientific Supplies
Supplier: United Scientific Supplies
BDPSC-AM-2OZ BDPSC-AM-1OZ BDPSC-AM-4OZ BDPSC-AM-8OZ BDPSC-AM-0.5OZ
470325-634PK 30.8 USD
470325-634 470325-636 470325-638 470325-640 470332-786
Bottles with Dropper, Boston Round, Amber, United Scientific Supplies
Bottles Dropping and Dispensing Bottles
Amber Boston round bottle with droppers for general use.
  • Screw cap with dropper assembly included
  • Non-porous glass is easy to clean and autoclavable
  • Amber color protects light-sensitive contents
These amber Boston round flint glass bottles include screw cap with dropper assembly are designed for general use. The glass is non-porous, which allows for easy cleaning or autoclaving. The amber color helps protect light sensitive materials.