Collapsible Ice Buckets, 5 Liters
Supplier: MTC BIO, INC.
B8300
470324-996EA 66 USD
470324-996
Collapsible Ice Buckets, 5 Liters
Ice Containers
This collapsible ice bucket is ideal for any lab looking to conserve storage space.
  • Inner-lid access port for better temperature retention
  • Attached handle for easy transport
  • Molded grip on base for pouring
  • Collapsible body saves storage space
BioBucket can be used as an ice bucket to keep samples cool over long periods of time, or as portable liquid container. The attached handle makes transportation a breeze while the molded base grip allows for easy pouring and cleaning. It is perfect for field use as an environmental sample collection and transport receptacle.